Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198962.3 | NP_945200.1 | 4 - 5 | 839 | 116 | |
GenBank mRNA | AF394597.1 | - | 3 - 4 | 468 | 116 | |
AK038551.1 | - | 4 - 5 | 840 | 116 | ||
AK048781.1 | - | 4 - 5 | 839 | 116 | ||
AY336084.1 | - | 4 - 5 | 807 | 116 | ||
AY336085.1 | - | 4 - 5 | 807 | 116 | ||
BC140957.1 | - | 4 - 5 | 835 | 9055569 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |