Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146046.1 | NP_001139518.1 | 8 - 9 | 1055 | 78 | |
NM_001146047.1 | NP_001139519.1 | 9 - 10 | 1054 | 78 | ||
NM_145616.4 | NP_663591.3 | 9 - 10 | 1253 | 78 | ||
GenBank mRNA | AK041627.1 | - | 7 - 8 | 757 | 78 | |
AK049268.1 | - | 9 - 10 | 1032 | 78 | ||
AK051325.1 | - | 8 - 9 | 860 | 78 | ||
AK082406.1 | - | 284 | 78 | |||
AK085022.1 | - | 1048 | 78 | |||
BC016574.1 | - | 8 - 9 | 822 | 3594577 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |