Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122873.1 | NP_001116345.1 | 1 - 2 | 71 | 93 | |
GenBank mRNA | BY230605.1 | - | 1 - 2 | 72 | 93 | |
CX203425.1 | - | 1 - 2 | 49 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |