Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oscp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172701.2 | NP_766289.2 | 3 - 4 | 543 | 87 | |
GenBank mRNA | AB154826.1 | - | 3 - 4 | 496 | 87 | |
AB286960.1 | - | 4 - 5 | 544 | 87 | ||
AK077467.1 | - | 3 - 4 | 509 | 87 | ||
AK077909.1 | - | 3 - 4 | 464 | 87 | ||
BC045150.1 | - | 3 - 4 | 489 | 6309923 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |