Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011986.4 | NP_036116.3 | 5 - 6 | 2041 | 95 | |
GenBank mRNA | AK002938.1 | - | 3 - 4 | 646 | 95 | |
AK154889.1 | - | 5 - 6 | 2022 | 95 | ||
AK170502.1 | - | 6 - 7 | 1756 | 95 | ||
BC017126.1 | - | 5 - 6 | 1972 | 4485839 | 95 | |
BZ690994.2 | - | 1 - 2 | 79 | 95 | ||
CD353011.1 | - | 1 - 2 | 73 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |