Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023603.3 | NP_076092.1 | 7 - 8 | 1905 | 70 | |
NR_045010.1 | 7 - 8 | 1905 | 70 | |||
GenBank mRNA | AK018483.1 | - | 2 - 3 | 198 | 70 | |
AK217233.1 | - | 1 - 2 | 67 | 70 | ||
AY034062.1 | - | 7 - 8 | 1820 | 70 | ||
BC037486.1 | - | 5 - 6 | 618 | 70 | ||
BC049227.1 | - | 4 - 5 | 476 | 70 | ||
BC055468.1 | - | 3 - 4 | 263 | 70 | ||
BC080848.1 | - | 7 - 8 | 1871 | 70 | ||
BC089305.1 | - | 7 - 8 | 1871 | 30360144 | 70 | |
CA319647.1 | - | 5 - 6 | 536 | 70 | ||
CJ304628.1 | - | 1 - 2 | 95 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |