Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027830.2 | NP_082106.2 | 11 - 12 | 2200 | 62 | |
GenBank mRNA | BC062185.1 | - | 2 - 3 | 277 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |