Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122892.1 | NP_001116364.1 | 5 - 6 | 882 | 61 | |
NM_001122893.1 | NP_001116365.1 | 6 - 7 | 952 | 61 | ||
NM_008054.2 | NP_032080.2 | 6 - 7 | 952 | 61 | ||
GenBank mRNA | AK156584.1 | - | 6 - 7 | 954 | 61 | |
AK171646.1 | - | 6 - 7 | 954 | 61 | ||
BC010554.1 | - | 5 - 6 | 565 | 61 | ||
BC032149.1 | - | 5 - 6 | 597 | 3602546 | 61 | |
BC092217.1 | - | 5 - 6 | 878 | 6403711 | 61 | |
M27266.1 | - | 7 - 8 | 664 | 61 | ||
U70324.1 | - | 6 - 7 | 665 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |