Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159544.1 | NP_001153016.1 | 3 - 4 | 1250 | 112 | |
NM_010237.3 | NP_034367.2 | 4 - 5 | 1094 | 112 | ||
GenBank mRNA | AK028855.1 | - | 4 - 5 | 1034 | 112 | |
AK052614.1 | - | 4 - 5 | 966 | 112 | ||
AK077264.1 | - | 3 - 4 | 1119 | 112 | ||
AK085191.1 | - | 3 - 4 | 1095 | 112 | ||
AK166379.1 | - | 4 - 5 | 1094 | 112 | ||
BC007137.1 | - | 4 - 5 | 1107 | 3601246 | 112 | |
L36132.1 | - | 3 - 4 | 1129 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |