Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030938.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030938.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030938.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030938.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030938.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030935.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030935.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030935.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030935.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030935.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030932.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030932.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030932.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030932.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030932.mm.1 |
NM_016799.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030936.mm.1 |
TC0400003760.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030936.mm.1 |
NM_001130477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030936.mm.1 |
NM_001130477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030936.mm.1 |
NM_016799.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030936.mm.1 |
NM_016799.3 |
Refseq |