Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001033257.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001033257.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195065.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195066.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195066.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195096.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
NM_001195096.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012877.mm.1 |
TC1000001768.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001033257.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001033257.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195065.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195065.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195066.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195066.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195096.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
NM_001195096.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012878.mm.1 |
TC1000001768.mm.1 |
MTA TC |