Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hey2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013904.1 | NP_038932.1 | 2 - 3 | 290 | 73 | |
GenBank mRNA | AB093589.1 | - | 2 - 3 | 158 | 73 | |
AF172287.1 | - | 2 - 3 | 290 | 73 | ||
AF173902.1 | - | 2 - 3 | 282 | 73 | ||
AF232240.1 | - | 2 - 3 | 286 | 73 | ||
AJ271867.1 | - | 2 - 3 | 290 | 73 | ||
AK031506.1 | - | 2 - 3 | 137 | 73 | ||
AK158000.1 | - | 2 - 3 | 291 | 73 | ||
BC103575.1 | - | 2 - 3 | 230 | 40045790 | 73 | |
BC103576.1 | - | 2 - 3 | 233 | 40045791 | 73 | |
BC105651.1 | - | 2 - 3 | 230 | 40046419 | 73 | |
BC105652.1 | - | 2 - 3 | 233 | 40046420 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |