Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pum1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159603.1 | NP_001153075.1 | 11 - 12 | 1762 | 68 | |
NM_001159604.1 | NP_001153076.1 | 11 - 12 | 1762 | 68 | ||
NM_001159605.1 | NP_001153077.1 | 9 - 10 | 1474 | 68 | ||
NM_001159606.1 | NP_001153078.1 | 7 - 8 | 1036 | 68 | ||
NM_030722.2 | NP_109647.2 | 11 - 12 | 1765 | 68 | ||
GenBank mRNA | AK122205.1 | - | 11 - 12 | 1745 | 68 | |
AK141921.1 | - | 11 - 12 | 1762 | 68 | ||
AK147185.1 | - | 7 - 8 | 1008 | 68 | ||
AK154059.1 | - | 10 - 11 | 1360 | 68 | ||
AK161135.1 | - | 9 - 10 | 1468 | 68 | ||
AK162742.1 | - | 11 - 12 | 1695 | 68 | ||
AK163977.1 | - | 11 - 12 | 1753 | 68 | ||
BC048174.1 | - | 11 - 12 | 1715 | 6825203 | 68 | |
BC050747.1 | - | 11 - 12 | 1735 | 6593374 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |