Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snhg3 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003270.2 | 1 - 2 | 91 | 84 | |||
GenBank mRNA | AJ006837.1 | - | LIT1313 U17HG |
1 - 2 | 38 | 84 | |
AK076188.1 | - | 1 - 2 | 39 | 84 | |||
BC100513.1 | - | 1 - 2 | 84 | 84 | |||
CN834544.1 | - | 1 - 2 | 94 | 84 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, H/ACA box 73b |
Snora73b | U17B | -137.0 | Sense | Chr. 4 - 132352322 - 132352526 on NCBI Build GRCm38 |
Downstream | phosphatase and actin regulator 4 | Phactr4 |
3110001B12Rik, AI527228, AW495572, C330013F19Rik, N28169, mKIAA4120 |
2.3kb | Sense | Chr. 4 - 132355922 - 132422446 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |