Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Asap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008232.2 | NP_001008233.1 | 12 - 13 | 1252 | 65 | |
GenBank mRNA | AK132294.1 | - | 12 - 13 | 1221 | 65 | |
BC085250.1 | - | 12 - 13 | 1205 | 30730459 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |