Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008232.2 | NP_001008233.1 | 22 - 23 | 2472 | 91 | |
GenBank mRNA | AK031005.1 | - | 2 - 3 | 303 | 91 | |
AK035099.1 | - | 8 - 9 | 923 | 91 | ||
AK132294.1 | - | 22 - 23 | 2441 | 91 | ||
BC085250.1 | - | 22 - 23 | 2425 | 30730459 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |