Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400031400.mm.1 |
TC0400003807.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031400.mm.1 |
NM_001126318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031400.mm.1 |
NM_001126318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031398.mm.1 |
TC0400003807.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031398.mm.1 |
NM_001126318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031398.mm.1 |
NM_001126318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031401.mm.1 |
TC0400003807.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031401.mm.1 |
NM_001126318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031401.mm.1 |
NM_001126318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031402.mm.1 |
TC0400003807.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031402.mm.1 |
NM_001126318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031402.mm.1 |
NM_001126318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031399.mm.1 |
TC0400003807.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031399.mm.1 |
NM_001126318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031399.mm.1 |
NM_001126318.1 |
Refseq |