Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111268.1 | NP_001104738.1 | 2 - 3 | 529 | 65 | |
NM_010349.2 | NP_034479.2 | 2 - 3 | 529 | 65 | ||
GenBank mRNA | AK141539.1 | - | 2 - 3 | 530 | 65 | |
BC060154.1 | - | 1 - 2 | 324 | 65 | ||
BC131639.1 | - | 1 - 2 | 210 | 65 | ||
BC131640.1 | - | 1 - 2 | 210 | 65 | ||
BC148311.2 | - | 1 - 2 | 210 | 65 | ||
D10054.1 | - | 2 - 3 | 442 | 65 | ||
X66117.1 | - | 1 - 2 | 266 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |