Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153574.2 | NP_705802.1 | 10 - 11 | 1256 | 89 | |
GenBank mRNA | AJ510136.1 | - | 10 - 11 | 1242 | 89 | |
AK036291.1 | - | 3 - 4 | 463 | 89 | ||
AK052421.1 | - | 10 - 11 | 1197 | 89 | ||
BC005554.1 | - | 10 - 11 | 1161 | 89 | ||
BC054053.1 | - | 10 - 11 | 1230 | 89 | ||
BC060978.1 | - | 10 - 11 | 1193 | 89 | ||
BC080861.1 | - | 10 - 11 | 1256 | 30357292 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |