Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011920.3 | NP_036050.1 | 9 - 10 | 1650 | 68 | |
GenBank mRNA | AF140218.1 | - | 8 - 9 | 1213 | 68 | |
BC053730.1 | - | 9 - 10 | 1662 | 30064535 | 68 | |
KF662864.1 | - | 9 - 10 | 1247 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |