Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253392.1 | NP_001240321.1 | 6 - 7 | 760 | 92 | |
NM_007958.1 | NP_031984.1 | 8 - 9 | 1221 | 92 | ||
GenBank mRNA | AK122454.1 | - | 8 - 9 | 1028 | 92 | |
AK134442.1 | - | 8 - 9 | 1072 | 92 | ||
AK147884.1 | - | 8 - 9 | 1010 | 92 | ||
BC042442.1 | - | 6 - 7 | 753 | 3980726 | 92 | |
X69942.1 | - | 8 - 9 | 1221 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |