Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK085078.1 | - | 310 | 68 | ||
AK157821.1 | - | 1 | 309 | 68 | ||
BC111036.1 | - | 1 | 282 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |