Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prdm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027547.2 | NP_081823.2 | 3 - 4 | 432 | 108 | |
GenBank mRNA | AK014501.1 | - | 3 - 4 | 356 | 108 | |
BC138901.1 | - | 3 - 4 | 406 | 8861923 | 108 | |
BC138902.1 | - | 3 - 4 | 406 | 8861924 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |