Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042743.2 | NP_001036208.1 | 19 - 20 | 2513 | 63 | |
NM_008641.3 | NP_032667.2 | 18 - 19 | 2492 | 63 | ||
GenBank mRNA | AB093264.1 | - | 9 - 10 | 1145 | 63 | |
BC060703.1 | - | 19 - 20 | 2083 | 6412811 | 63 | |
CK633885.1 | - | 4 - 5 | 541 | 63 | ||
U02313.1 | - | 18 - 19 | 2162 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |