Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178058.1 | NP_001171529.1 | 4 - 5 | 693 | 116 | |
NM_001178059.1 | NP_001171530.1 | 4 - 5 | 693 | 116 | ||
NM_001178060.1 | NP_001171531.1 | 4 - 5 | 693 | 116 | ||
NM_153162.3 | NP_694802.2 | 4 - 5 | 693 | 116 | ||
GenBank mRNA | AK012699.1 | - | 4 - 5 | 686 | 116 | |
AK041046.1 | - | 4 - 5 | 694 | 116 | ||
AK080362.1 | - | 4 - 5 | 684 | 116 | ||
AK134987.1 | - | 4 - 5 | 684 | 116 | ||
BC076605.1 | - | 4 - 5 | 651 | 30628949 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |