Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008636.4 | NP_032662.3 | 4 - 5 | 919 | 75 | |
GenBank mRNA | AK150295.1 | - | 5 - 6 | 981 | 75 | |
AK182707.1 | - | 1 - 2 | 94 | 75 | ||
AY260849.1 | - | 3 - 4 | 773 | 75 | ||
AY260850.1 | - | 3 - 4 | 773 | 75 | ||
BC017679.1 | - | 4 - 5 | 881 | 4210829 | 75 | |
X71327.1 | - | 4 - 5 | 869 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |