Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smyd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160127.1 | NP_001153599.1 | 1 - 2 | 212 | 86 | |
NM_009762.2 | NP_033892.2 | 1 - 2 | 212 | 86 | ||
GenBank mRNA | AK076266.1 | - | 1 - 2 | 176 | 86 | |
AK142153.1 | - | 1 - 2 | 180 | 86 | ||
AK142252.1 | - | 1 - 2 | 190 | 86 | ||
BC076601.1 | - | 1 - 2 | 190 | 30609878 | 86 | |
BY089666.1 | - | 1 - 2 | 213 | 86 | ||
U76373.2 | - | 1 - 2 | 184 | 86 | ||
U76374.2 | - | 1 - 2 | 184 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |