Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thnsl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033929.2 | NP_001029101.1 | 2 - 3 | 326 | 78 | |
NM_178413.5 | NP_848500.3 | 3 - 4 | 394 | 78 | ||
GenBank mRNA | AK165463.1 | - | 2 - 3 | 292 | 78 | |
BC025604.1 | - | 3 - 4 | 357 | 5250319 | 78 | |
BC051244.1 | - | 2 - 3 | 288 | 4975661 | 78 | |
CA491740.1 | - | 2 - 3 | 359 | 78 | ||
CB600010.1 | - | 3 - 4 | 364 | 78 | ||
CX223060.1 | - | 2 - 3 | 326 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |