Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026560.4 | NP_080836.3 | 7 - 8 | 848 | 57 | |
GenBank mRNA | AK076065.1 | - | 7 - 8 | 805 | 57 | |
AK076422.1 | - | 7 - 8 | 838 | 57 | ||
AK077717.1 | - | 7 - 8 | 801 | 57 | ||
AK077920.1 | - | 7 - 8 | 771 | 57 | ||
AK078404.1 | - | 7 - 8 | 801 | 57 | ||
AK082800.1 | - | 7 - 8 | 813 | 57 | ||
AY550907.1 | - | 7 - 8 | 838 | 57 | ||
BC068181.1 | - | 6 - 7 | 762 | 30634239 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |