Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF077330.1 | - | 5 - 6 | 405 | 123 | |
AK014433.1 | - | 6 - 7 | 123 | |||
AK032514.1 | - | 6 - 7 | 639 | 123 | ||
AK048148.1 | - | 6 - 7 | 437 | 123 | ||
AK159381.1 | - | 6 - 7 | 525 | 123 | ||
AK166655.1 | - | 5 - 6 | 395 | 123 | ||
AK168859.1 | - | 6 - 7 | 440 | 123 | ||
AY029181.1 | - | 5 - 6 | 405 | 123 | ||
BC080776.1 | - | 5 - 6 | 358 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |