Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081111.2 | NP_001074580.1 | 10 - 11 | 2494 | 66 | |
GenBank mRNA | BC150931.1 | - | 10 - 11 | 2505 | 9087842 | 66 |
BE627586.1 | - | 2 - 3 | 244 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |