Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uroc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144940.2 | NP_659189.2 | 3 - 4 | 377 | 76 | |
GenBank mRNA | AK149464.1 | - | 3 - 4 | 378 | 76 | |
BC022133.1 | - | 3 - 4 | 360 | 5098655 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |