Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021385.2 | NP_067360.2 | 9 - 10 | 1106 | 140 | |
GenBank mRNA | AF205278.1 | - | 9 - 10 | 1030 | 140 | |
AK012795.1 | - | 9 - 10 | 1107 | 140 | ||
AK028505.1 | - | 9 - 10 | 1026 | 140 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |