Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080448.4 | NP_536696.4 | 20 - 21 | 3331 | 108 | |
GenBank mRNA | AF338472.1 | - | 19 - 20 | 2489 | 108 | |
AF496547.1 | - | 20 - 21 | 2561 | 108 | ||
CF744803.1 | - | 1 - 2 | 123 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Mus musculus Probe spans exons |