Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138592.4 | NP_613058.1 | 2 - 3 | 360 | 66 | |
GenBank mRNA | AK010443.1 | - | 2 - 3 | 370 | 66 | |
AK033105.1 | - | 2 - 3 | 357 | 66 | ||
AK045062.1 | - | 2 - 3 | 357 | 66 | ||
AK088354.1 | - | 2 - 3 | 372 | 66 | ||
AK165625.1 | - | 2 - 3 | 341 | 66 | ||
AK167668.1 | - | 2 - 3 | 348 | 66 | ||
AK168003.1 | - | 2 - 3 | 367 | 66 | ||
AK201881.1 | - | 2 - 3 | 127 | 66 | ||
BC026983.1 | - | 2 - 3 | 335 | 5365862 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |