Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138592.4 | NP_613058.1 | 6 - 7 | 971 | 73 | |
GenBank mRNA | AK010443.1 | - | 6 - 7 | 981 | 73 | |
AK018595.1 | - | 6 - 7 | 972 | 73 | ||
AK033105.1 | - | 6 - 7 | 968 | 73 | ||
AK045062.1 | - | 6 - 7 | 968 | 73 | ||
AK088354.1 | - | 6 - 7 | 983 | 73 | ||
AK165625.1 | - | 6 - 7 | 952 | 73 | ||
AK167668.1 | - | 6 - 7 | 959 | 73 | ||
AK168003.1 | - | 6 - 7 | 978 | 73 | ||
AK182709.1 | - | 1 - 2 | 153 | 73 | ||
BC026983.1 | - | 6 - 7 | 946 | 5365862 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |