Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016886.1 | - | 8 - 9 | 570 | 63 | |
BC100525.1 | - | 8 - 9 | 527 | 30920200 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |