Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007657.3 | NP_031683.1 | 6 - 7 | 653 | 80 | |
GenBank mRNA | AK002251.1 | - | 6 - 7 | 659 | 80 | |
AK012793.1 | - | 6 - 7 | 659 | 80 | ||
AK151443.1 | - | 6 - 7 | 619 | 80 | ||
AK151619.1 | - | 6 - 7 | 606 | 80 | ||
AK170310.1 | - | 6 - 7 | 606 | 80 | ||
BC070474.1 | - | 6 - 7 | 653 | 30634238 | 80 | |
L08115.1 | - | 6 - 7 | 591 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |