Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138677.2 | NP_619618.1 | 2 - 3 | 657 | 112 | |
GenBank mRNA | AB042828.1 | - | 2 - 3 | 646 | 112 | |
AK030943.1 | - | 2 - 3 | 658 | 112 | ||
AK037889.1 | - | 2 - 3 | 220 | 112 | ||
AK082920.1 | - | 2 - 3 | 610 | 112 | ||
AK129090.1 | - | 2 - 3 | 624 | 112 | ||
AK151668.1 | - | 2 - 3 | 632 | 112 | ||
AK152724.1 | - | 2 - 3 | 633 | 112 | ||
AK152979.1 | - | 2 - 3 | 112 | |||
AK188251.1 | - | 1 - 2 | 73 | 112 | ||
BC023237.1 | - | 2 - 3 | 605 | 2654113 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |