Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600010802.mm.1 |
TC0600001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010802.mm.1 |
NM_001081048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010802.mm.1 |
NM_001081048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010803.mm.1 |
TC0600001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010803.mm.1 |
NM_001081048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010803.mm.1 |
NM_001081048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010800.mm.1 |
TC0600001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010800.mm.1 |
NM_001081048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010800.mm.1 |
NM_001081048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010804.mm.1 |
TC0600001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010804.mm.1 |
NM_001081048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010804.mm.1 |
NM_001081048.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010801.mm.1 |
TC0600001354.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600010801.mm.1 |
NM_001081048.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600010801.mm.1 |
NM_001081048.2 |
Refseq |