Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 13 - 14 | 2289 | 76 | |
GenBank mRNA | AK144877.1 | - | 13 - 14 | 2260 | 76 | |
AK159079.1 | - | 13 - 14 | 2266 | 76 | ||
AK166055.1 | - | 13 - 14 | 2270 | 76 | ||
BC011506.3 | - | 13 - 14 | 2223 | 76 | ||
BC080691.1 | - | 13 - 14 | 1975 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |