Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145997.2 | NP_666109.2 | 17 - 18 | 2943 | 70 | |
GenBank mRNA | AK052087.1 | - | 1 - 2 | 924 | 70 | |
AK135085.1 | - | 2 - 3 | 271 | 70 | ||
AK144877.1 | - | 17 - 18 | 2914 | 70 | ||
AK166055.1 | - | 17 - 18 | 2924 | 70 | ||
BC011506.3 | - | 17 - 18 | 2877 | 70 | ||
BC022581.1 | - | 2 - 3 | 218 | 70 | ||
BC080691.1 | - | 17 - 18 | 2629 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |