Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010654.3 | NP_034784.1 | 2 - 3 | 111 | 124 | |
GenBank mRNA | AF030311.1 | - | 1 - 2 | 91 | 124 | |
AF030312.1 | - | 2 - 3 | 140 | 124 | ||
AF039025.1 | - | 2 - 3 | 105 | 124 | ||
AF057714.1 | - | 1 - 2 | 91 | 124 | ||
AK136548.1 | - | 2 - 3 | 112 | 124 | ||
AK162976.1 | - | 2 - 3 | 113 | 124 | ||
BC117112.1 | - | 2 - 3 | 129 | 40126431 | 124 | |
BC120854.1 | - | 2 - 3 | 131 | 40129777 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |