Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009531.2 | NP_033557.2 | 5 - 6 | 693 | 107 | |
GenBank mRNA | AB071144.1 | - | 5 - 6 | 630 | 107 | |
AK004713.1 | - | 5 - 6 | 633 | 107 | ||
AK028595.1 | - | 5 - 6 | 631 | 107 | ||
AK166981.1 | - | 5 - 6 | 628 | 107 | ||
AK170975.1 | - | 5 - 6 | 625 | 107 | ||
BC052725.1 | - | 5 - 6 | 655 | 6812198 | 107 | |
U27398.1 | - | 5 - 6 | 601 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |