Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 9 - 10 | 2409 | 56 | |
NM_007992.2 | NP_032018.2 | 10 - 11 | 2550 | 56 | ||
GenBank mRNA | AK159361.1 | - | 9 - 10 | 2721 | 56 | |
AK161522.1 | - | 9 - 10 | 2352 | 56 | ||
AK168688.1 | - | 9 - 10 | 2347 | 56 | ||
BC005443.1 | - | 9 - 10 | 2389 | 3490759 | 56 | |
X75285.1 | - | 10 - 11 | 2450 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |