Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159507.1 | NP_001152979.1 | 7 - 8 | 799 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |