Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
TC0600000987.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_001164078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_001164078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_001164079.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_001164079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_011585.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007625.mm.1 |
NM_011585.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
TC0600000987.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_001164078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_001164078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_001164079.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_001164079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_011585.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007620.mm.1 |
NM_011585.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
TC0600000987.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_001164078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_001164078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_001164079.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_001164079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_011585.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007618.mm.1 |
NM_011585.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
TC0600000987.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_001164078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_001164078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_001164079.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_001164079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_011585.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007619.mm.1 |
NM_011585.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
TC0600000987.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_001164078.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_001164078.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_001164079.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_001164079.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_011585.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600007609.mm.1 |
NM_011585.4 |
Refseq |