Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624873 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624872 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624897 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624896 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624871 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594079 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
7003 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
497044 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758521 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
605307 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
764171 |
BC002253.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
764171 |
NM_133839.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
197326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
341242 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
245201 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
504385 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
16120 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P162660 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P126979 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624868 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624900 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624894 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624891 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624863 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624892 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624889 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624879 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624873 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624867 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624865 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624887 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624897 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624869 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624888 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624857 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624881 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624859 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624880 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624875 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624878 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624886 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624860 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624862 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624877 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624883 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624861 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624899 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P24624871 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P126979 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01380009 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P162660 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
D86725.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
CK378639.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
AK088156.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P158210 |
AF004105.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P129945 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P162660 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P126979 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05079306 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05746285 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05488711 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05488708 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05667089 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05746281 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05488707 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P05079302 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624871 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624897 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624872 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624873 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P24624896 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624875 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624848 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624871 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624712 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624759 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624782 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624883 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624708 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624908 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624769 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624806 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624843 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624808 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624865 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624778 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624838 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624859 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624739 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624761 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624863 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624747 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624821 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624730 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624881 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624783 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624841 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624926 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624758 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624860 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624746 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624805 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624880 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624846 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624831 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624900 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624897 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624727 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624890 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624775 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624781 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624788 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624773 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624741 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624873 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624813 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624811 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624718 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624931 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624787 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624867 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624934 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624731 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624923 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624765 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624922 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624862 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624855 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624911 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624925 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624799 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624920 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624817 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624916 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624823 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624785 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624888 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624780 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624762 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624891 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624797 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624793 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624907 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624820 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624945 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624928 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624744 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624903 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624790 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624802 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624918 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624861 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624714 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624768 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624737 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624722 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624857 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624919 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624733 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624868 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624839 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624795 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624932 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624902 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624828 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624899 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624940 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624836 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624771 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624886 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624803 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624853 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624832 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624825 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624749 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624936 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624892 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624914 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624735 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624878 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624835 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624827 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624829 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624725 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624879 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624809 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P24624743 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05862123 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624873 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05362142 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624831 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624838 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624856 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624836 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624850 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624846 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05362137 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624823 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624879 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05540484 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624886 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624828 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624848 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624829 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624881 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624859 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624867 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624841 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05862126 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624863 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624844 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05487287 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05540489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624845 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05487288 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624871 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624825 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624827 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624861 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624875 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624880 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05347127 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624862 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624883 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624857 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05362131 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624869 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624878 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P24624855 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P05488710 |
- |
- |