Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12rb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008354.3 | NP_032380.1 | 6 - 7 | 991 | 95 | |
GenBank mRNA | BC137745.1 | - | 5 - 6 | 903 | 8860764 | 95 |
U64199.1 | - | 6 - 7 | 992 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |