Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025467.1 | NP_079743.1 | 3 - 4 | 238 | 94 | |
GenBank mRNA | AK007451.1 | - | 3 - 4 | 238 | 94 | |
AK007705.1 | - | 3 - 4 | 238 | 94 | ||
AK008692.1 | - | 3 - 4 | 239 | 94 | ||
AK008986.1 | - | 3 - 4 | 239 | 94 | ||
AY943909.1 | - | 3 - 4 | 246 | 94 | ||
BC099445.1 | - | 3 - 4 | 227 | 30788014 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |